Learn More
7-Bromo-5-fluorobenzo[b]furan, 97%
Product Code. 15426837
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15426837
Brand: Thermo Scientific Alfa Aesar H33948.03
Request Bulk or Custom Format
CAS: 253429-19-5 | C8H4BrFO | 215.021 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 253429-19-5
Molecular Formula C8H4BrFO
Molecular Weight (g/mol) 215.021
MDL Number MFCD09056780
InChI Key TYBMMMOEVBNTDU-UHFFFAOYSA-N
PubChem CID 22144816
IUPAC Name 7-bromo-5-fluoro-1-benzofuran
SMILES C1=COC2=C(C=C(C=C21)F)Br
Specifications
Quantity 1 g
Synonym 7-bromo-5-fluorobenzofuran, 7-bromo-5-fluorobenzo b furan, 5-fluoro-7-bromobenzofuran, 7-bromo-5-fluoro-benzofuran, benzofuran,7-bromo-5-fluoro, benzofuran, 7-bromo-5-fluoro, 7-bromanyl-5-fluoranyl-1-benzofuran
Formula Weight 215.02
Percent Purity 97%
Chemical Name or Material 7-Bromo-5-fluorobenzo[b]furan