Learn More
7-Amino-3-(1-pyridylmethyl)-3-cephem-4-carboxylic Acid Hydroiodide, TRC
Specifications
Specifications
| Chemical Name or Material | (6R,7R)-7-Amino-8-oxo-3-[(pyridin-1-ium-1-yl)methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate Hydroiodide |
| CAS | 100988-63-4 |
| Molecular Formula | C13H14IN3O3S |
| Quantity | 100 mg |
| Synonym | 1-[[(6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-pyridinium Inner Salt Hydroiodide, (6R-trans)-1-[(7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo |
| InChI Formula | InChI=1S/C13H13N3O3S.HI/c14-9-11(17)16-10(13(18)19)8(7-20-12(9)16)6-15-4-2-1-3-5-15;/h1-5,9,12H,6-7,14H2;1H/t9-,12-;/m1./s1 |
| SMILES | [H][C@@]1([C@]([H])(N)C2=O)N2C(C(O)=O)=C(C[N+]3=CC=CC=C3)CS1.[I-] |
| IUPAC Name | 1-(((6R,7R)-7-amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)pyridin-1-ium iodide |
| Molecular Weight (g/mol) | 419.24 |
| Formula Weight | 418.98 |