Learn More
Viking Optical™ 6X Linen Tester Magnifier
Product Code. p-8011081
Encompass_Preferred
Change view
Click to view available options
Description:
25.4 x 3.2 mm
Scale: 25 x 1 mm
Unit Size:
Each
Product Code. Description unitSize
12836335 Scale: 25 x 1 mm Each
12846335 25.4 x 3.2 mm Each
2 options
Product Code. 12836335
Brand: Viking Optical™ 5055
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Two element glass lens in a black finished metal folding mount. 6x magnification.
Specifications
Description Scale: 25 x 1 mm
Magnification 6x