missing translation for 'onlineSavingsMsg'
Learn More

(6R)-5,6,7,8-Tetrahydro-L-Biopterin Dihydrochloride , MP Biomedical

Product Code. 11426145 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426145 50 mg 50mg
11416145 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426145 Supplier MP Biomedicals Supplier No. 0215362250
  • ONLINE EXCLUSIVE PRICE
    7385.00 NOK
    7034.76 NOK / 50mg
    Save 350.24 NOK
    4% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is a co-factor for nitric oxide synthase.

  • (6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is necessary for vivo for the conversion of phenylalanine to tyrosine and for the production of the hormone epinephrine and of the monoamine neurotransmitters, serotonin, dopamine, and norepinephrine
  • It is also involved in apoptosis and other cellular events mediated by nitric oxide production and catalyzes the cleavage of alkylglycerol ethers
  • (6R)-BH4 has been used as a buffer component for assaying tyrosine hydroxylase enzyme activity in mouse brain tissues
  • Natural cofactor for phenylalanine hydroxylase, tyrosine hydroxylase, tryptophan hydroxylase, and nitric oxide synthetase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H17Cl2N5O3
Molecular Weight (g/mol) 314.17
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, biopten, kuvan, sapropterin hydrochloride, 6r-5,6,7,8-tetrahydro-l-biopterin dihydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl
PubChem CID 636369
ChEBI CHEBI:32120

Specifications

Color White
Packaging Amber Glass Bottle
Quantity 50 mg
Percent Purity 99.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.