Learn More
Masterflex™ 65mm Correlated Flowmeters with Valve
Specifications
Specifications
| Type | Variable Area Flowmeter |
| Accuracy | ±2% (Full Scale) |
| Airflow Rate | 42.094 LPM |
| Float Material | Stainless Steel (316) |
| Pressure Rating | 200 psig |
| Connections | 1/8 in. NPT (F) |
| Flow Rate Range | 35,100 mL/min. CO2, 40,053 mL/min. O2, 43,487 mL/min. N2, 37,441 mL/min. Ar, 154,750 mL/min. H2 |
| For Use With (Application) | Ideal for tight measuring areas |
| Length (Metric) | 65 mm |
| Repeatability | ±0.25% |
| Show More |