Learn More
Hamilton™ 600 Microliter Syringes, No Needle Included
Product Code. 11532505
Change view
Click to view available options
Volume (Metric):
2.5 μL
5 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
11532505 5 μL Each
11522505 2.5 μL Each
2 options
Product Code. 11532505
Brand: Hamilton™ 763301
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0.25 μL up to 5 μL. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.
  • Half stroke version of the original icroliter 700 series syringes
  • Ideal for dispensing volumes from 0.25 μL up to 5 μL
  • Often used in applications that require one handed operation because of the short stroke length and the reinforced plunger
Specifications
Needle Gauge 22s G
Volume (Metric) 5 μL