Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Frontier™ R-A12x2/18MI Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
1 x 1.5 mL
Unit Size:
Pack of 6
Description
- Allows you to centrifuge tubes that are smaller than the space provided in the rotor carrier
- For over 16000 x G
- Suitable for use in Frontier™ R-A12x2/18MI Rotors
Specifications
Specifications
| Capacity | 1 x 1.5 mL |
| For Use With (Equipment) | R-A12x2/18MI Rotor |
| Description | Adapter, 1x1.5ml for over16000xg, 6/pk |
| No. of Places | 1 |
| No. per Pack | 2 |