missing translation for 'onlineSavingsMsg'
Learn More

6-Methyluracil, 97%

Product Code. 11481227
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11481227

Brand: Thermo Scientific Alfa Aesar B24191.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-48-2 | C5H6N2O2 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-48-2
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006028
InChI Key SHVCSCWHWMSGTE-UHFFFAOYSA-N
Synonym 6-methyluracil, 2,4-dihydroxy-6-methylpyrimidine, pseudothymine, 6-methylpyrimidine-2,4 1h,3h-dione, 4-methyluracil, 6-methylpyrimidine-2,4-diol, 2,4 1h,3h-pyrimidinedione, 6-methyl, uracil, 6-methyl, 6-methyl-2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 6-methyl
PubChem CID 12283
ChEBI CHEBI:74733
IUPAC Name 6-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CC(=O)NC(=O)N1

Specifications

Melting Point 317°C to 320°C
Quantity 500 g
Beilstein 115647
Merck Index 14,6133
Formula Weight 126.12
Percent Purity 97%
Chemical Name or Material 6-Methyluracil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.