missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, 98%, Thermo Scientific Chemicals

Catalog Number p-7026091
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
91-62-3
C10H9N
143.19
MFCD00006804
LUYISICIYVKBTA-UHFFFAOYSA-N
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
7059
6-methylquinoline
CC1=CC=C2N=CC=CC2=C1
This item is not returnable. View return policy
91-62-3
C10H9N
143.19
MFCD00006804
LUYISICIYVKBTA-UHFFFAOYSA-N
quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
7059
6-methylquinoline
CC1=CC=C2N=CC=CC2=C1

6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point -22°C
Density 1.066
Boiling Point 257°C to 259°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 5 g
Beilstein 110336
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 6-Methylquinoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'