Learn More
6-Methylquinoline, 98%
Product Code. 11354639
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11354639 5 g 5g
11404967 25 g 25g
11414967 100 g 100g
3 options
Product Code. 11354639
Brand: Thermo Scientific Alfa Aesar B21892.06
Request Bulk or Custom Format
CAS: 91-62-3 | C10H9N | 143.19 g/mol
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
Solubility
Not miscible or difficult to mix with water.
Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91-62-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006804
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
PubChem CID 7059
IUPAC Name 6-methylquinoline
SMILES CC1=CC=C2N=CC=CC2=C1
Specifications
Melting Point -22°C
Density 1.066
Boiling Point 257°C to 259°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 5 g
Beilstein 110336
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 6-Methylquinoline