missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Use as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Use as primary and secondary intermediate.
Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
Use as primary and secondary intermediate.
Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
Specifications
Melting Point | 32°C to 34°C |
Boiling Point | 105°C to 108°C (12 mmHg) |
Flash Point | 109°C (228°F) |
Quantity | 1 g |
Solubility Information | Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene. |
Formula Weight | 123.15 |
Percent Purity | 98% |
Chemical Name or Material | 6-Methyl-2-pyridinemethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'