missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals

Catalog Number 11306899
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1
This item is not returnable. View return policy
1122-71-0
C7H9NO
123.16
MFCD00023520
JLVBSBMJQUMAMW-UHFFFAOYSA-N
6-methyl-2-pyridinemethanol, 6-methylpyridin-2-yl methanol, 2-pyridinemethanol, 6-methyl, 6-methyl-2-pyridylmethanol, 6-methylpyridine-2-methanol, 2-hydroxymethyl-6-methylpyridine, 6-methyl-2-pyridyl methan-1-ol, pubchem20959, acmc-2099dw, 6-methyl-2-pyridylcarbinol
70736
(6-methylpyridin-2-yl)methanol
CC1=CC=CC(CO)=N1

Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
TRUSTED_SUSTAINABILITY
Melting Point 32°C to 34°C
Boiling Point 105°C to 108°C (12 mmHg)
Flash Point 109°C (228°F)
Quantity 1 g
Solubility Information Low soluble in water,high soluble in ethanol,acetic acid. Soluble in benzene,toluene.
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-pyridinemethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'