Learn More
6-Methyl-1,3,5-triazine-2,4(1H,3H)-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Methyl-1,3,5-triazine-2,4(1H,3H)-dione |
| CAS | 933-19-7 |
| Molecular Formula | C4 H5 N3 O2 |
| Quantity | 10 g |
| Synonym | 1,3,5-Triazin-2(1H)-one, 4-hydroxy-6-methyl-, 1,3,5-Triazine-2,4(1H,3H)-dione, 6-methyl- (9CI), s-Triazine-2,4(1H,3H)-dione, 6-methyl- (8CI), s-Triazine-2,4-diol, 6-methyl- (6CI,7CI), 4-Hydroxy-6-methyl-1,3,5-triazin-2(1H)-one, 2,4-Dihydroxy-6-methyl-1,3,5-triazine, 2,4-Dihydroxy-6-methyl-s-triazine, 2,4-Dioxo-6-methyltetrahydro-s-triazine, 2-Methyl-4,6-dihydroxy-s-triazine, 6-Methyl-1,3,5-triazine-2,4(1H,3H)-dione, 6-Methyl-1H,3H-s-triazine-2,4-dione |
| InChI Formula | InChI=1S/C4H5N3O2/c1-2-5-3(8)7-4(9)6-2/h1H3,(H2,5,6,7,8,9) |
| SMILES | CC1=NC(=O)NC(=O)N1 |
| IUPAC Name | 6-methyl-1H-1,3,5-triazine-2,4-dione |
| Molecular Weight (g/mol) | 127.1 |
| Formula Weight | 127.0382 |