Learn More
6-Methoxy-3,4-dihydronaphthalen-1(2H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1078-19-9 |
| Molecular Formula | C11H12O2 |
| MDL Number | MFCD00001695 |
| Quantity | 25 g |
| SMILES | O=C1CCCC2=C1C=CC(OC)=C2 |
| Molecular Weight (g/mol) | 176.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one |