Learn More
6-Methoxy-2-naphthylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Product Code. 10311770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311770 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10311770 Supplier Thermo Scientific Acros Supplier No. 431800500
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 38046-82-1 | C11H9BrMgO | 261.40 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 38046-82-1, 109-99-9
CAS Min % 10.82, 83.76
CAS Max % 16.24, 89.18
Color Brown
Density 0.9660g/mL
Flash Point −25°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C11H9BrMgO
MDL Number MFCD01311488
Quantity 50 mL
Specific Gravity 0.966
Synonym 6-methoxy-2-naphthylmagnesium bromide, sogqgmfueumyjd-uhfffaoysa-m, 6-methoxynaphthalen-2-ylmagnesium bromide, 4-fluorobenzylmagnesium chloride 0.25m, 6-methoxy-2-naphthylmagnesium bromide, 0.5m in thf, 6-methoxy-2-naphthylmagnesium bromide 0.5 m in tetrahydrofuran, 6-methoxy-2-naphthylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key SOGQGMFUEUMYJD-UHFFFAOYSA-M
SMILES COC1=CC2=CC=C([Mg]Br)C=C2C=C1
IUPAC Name bromo(6-methoxynaphthalen-2-yl)magnesium
Molecular Weight (g/mol) 261.40
PubChem CID 11242453
Formula Weight 261.4
Physical Form Liquid
Chemical Name or Material 6-Methoxy-2-naphthylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 6-Methoxy-2-naphthylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.