Learn More
6-Methoxy-1,2,3,4-tetrahydroisoquinoline, TRC
Product Code. 30473819
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473819 250 mg 250mg
30476114 50 mg 50mg
30481983 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30473819 Supplier Toronto Research Chemicals Supplier No. TRCM268675250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42923-77-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
Synonym 1,2,3,4-Tetrahydro-6-methoxyisoquinoline, 6-Methoxy-1,2,3,4-tetrahydroisoquinoline
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroisoquinoline
SMILES COC1=CC=C(CNCC2)C2=C1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C10H13NO/c1-12-10-3-2-9-7-11-5-4-8(9)6-10/h2-3,6,11H,4-5,7H2,1H3
Formula Weight 163.1
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroisoquinoline