Learn More
6-Heptyn-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Heptyn-1-ol |
| CAS | 63478-76-2 |
| Molecular Formula | C7H12O |
| Quantity | 250 mg |
| Synonym | 7-Hydroxyhept-1-yne |
| InChI Formula | InChI=1S/C7H12O/c1-2-3-4-5-6-7-8/h1,8H,3-7H2 |
| SMILES | OCCCCCC#C |
| IUPAC Name | hept-6-yn-1-ol |
| Molecular Weight (g/mol) | 112.17 |
| Formula Weight | 112.0888 |