Learn More
6-Heptenenitrile, 96%
Product Code. 11317926
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317926 5 g 5g
11307926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11317926 Supplier Thermo Scientific Alfa Aesar Supplier No. L13137.06
 Restricted Item
In Stock
Add to basket
CAS: 5048-25-9 | C7H11N | 109.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5048-25-9
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.17
MDL Number MFCD00151804
InChI Key ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
Synonym 6-heptenenitrile, 6-cyano-1-hexene, acmc-20alyq
PubChem CID 4140856
IUPAC Name hept-6-enenitrile
SMILES C=CCCCCC#N
Specifications
Boiling Point 75°C to 78°C (13 mmHg)
Flash Point >100°C (212°F)
Linear Formula CH2=CH(CH2)4CN
Refractive Index 1.433
Quantity 5 g
UN Number UN3276
Beilstein 1740406
Formula Weight 109.17
Percent Purity 97%
Chemical Name or Material 6-Heptenenitrile