Learn More
Justrite™ 6 Gallon Drum
Product Code. 15143416
Click to view available options
Capacity (English):
6 gal.
Unit Size:
Each
Product Code. 15143416
Brand: Justrite™ 26050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Justrite™ 6 Gallon Drum for use with cease-fire parts.
  • Drum only
  • Holds up to 6 gallons
Specifications
Capacity (English) 6 gal.
Includes Drum Only
Height (Metric) 40 cm
Diameter (Metric) Outer 29.8 mm