Learn More
6-Formyl-2-(methylsulfanyl)nicotinonitrile, 97%, Thermo Scientific™
Product Code. 10039531
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10039531 250 mg Amber Glass Bottle 250mg
10495522 1 g Amber Glass Bottle 1g
10103624 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10039531 Supplier Thermo Scientific Maybridge Supplier No. SPB04394CB
 Restricted Item
Estimated Shipment: 26-10-2026
Chemical Identifiers
CAS 175277-27-7
Molecular Formula C8H6N2OS
Molecular Weight (g/mol) 178.21
MDL Number MFCD00173824
InChI Key UBGVOMZGBKLUOU-UHFFFAOYSA-N
Synonym 6-formyl-2-methylsulfanyl nicotinonitrile, 3-cyano-2-methylthio pyridine-6-carboxaldehyde, 6-formyl-2-methylsulfanyl pyridine-3-carbonitrile, 6-formyl-2-methylthio nicotinonitrile, 6-formyl-2-methylthio pyridine-3-carbonitrile, 6-formyl-2-methylthio-3-pyridinecarbonitrile, 3-pyridinecarbonitrile,6-formyl-2-methylthio, 6-formyl-2-methylsulphanyl nicotinonitrile, 6-formyl-2-methylthiopyridine-3-carbonitrile, 6-formyl-2-methylsulfanyl-pyridine-3-carbonitrile
PubChem CID 2799504
IUPAC Name 6-formyl-2-methylsulfanylpyridine-3-carbonitrile
SMILES CSC1=C(C=CC(C=O)=N1)C#N
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 6-Formyl-2-(methylsulfanyl)nicotinonitrile