missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1,2,3,4-tetrahydro-naphthalene-2-carboxylic Acid, TRC

Product Code. 30406678
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406678 50 mg
30407501 10 mg
30393299 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406678 Supplier Toronto Research Chemicals Supplier No. TRCF60072350MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885269-59-0
Molecular Formula C11 H11 F O2
Molecular Weight (g/mol) 194.2
IUPAC Name 6-fluorotetralin-2-carboxylic acid
SMILES OC(=O)C1CCc2cc(F)ccc2C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11FO2/c12-10-4-3-7-5-9(11(13)14)2-1-8(7)6-10/h3-4,6,9H,1-2,5H2,(H,13,14)
Formula Weight 194.0743
Chemical Name or Material 6-Fluoro-1,2,3,4-tetrahydro-naphthalene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.