Learn More
6-Chloro-2H-chromene-3-carboxylic acid, 97%, Thermo Scientific™
Product Code. 11475463
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475463 5 g 5g
11465463 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11475463 Supplier Thermo Scientific Alfa Aesar Supplier No. A13425.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83823-06-7
Molecular Formula C10H7ClO3
Molecular Weight (g/mol) 210.613
MDL Number MFCD00052362
InChI Key ZRCGKWSNRRTAJY-UHFFFAOYSA-N
Synonym 6-chloro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid, 6-chloro, 2h-1-benzopyran-3-carboxylic acid, 6-chloro, maybridge1_005975, acmc-20alzg, chlorochromenecarboxylicacid, #, 6-chloro 2h-benzopyran-3-carboxylic acid, 6-chloro 2h-1-benzopyran-3-carboxylic acid, 6-chloro-2h-1-benzopyran-3-carboxylic
PubChem CID 596929
IUPAC Name 6-chloro-2H-chromene-3-carboxylic acid
SMILES C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)O
Specifications
Melting Point 240°C to 246°C
Quantity 5 g
Beilstein 4426673
Formula Weight 210.62
Percent Purity 97%
Chemical Name or Material 6-Chloro-2H-chromene-3-carboxylic acid