missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-fluoro-3-methylbenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 11490977
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261762-81-6
C8H5Cl2FO
207.025
MFCD01631371
NWTPJKCARLIKFP-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methylbenzoylchloride, acmc-20alzl, 2-fluoro-6-chloro-3-methyl-benzoyl chloride, 6-chloro-2-fluoro-3-methyl-benzoyl chloride, benzoyl chloride,2-chloro-6-fluoro-3-methyl, 6-chloranyl-2-fluoranyl-3-methyl-benzoyl chloride, benzoyl chloride, 6-chloro-2-fluoro-3-methyl-9ci
2773671
6-chloro-2-fluoro-3-methylbenzoyl chloride
CC1=C(C(=C(C=C1)Cl)C(=O)Cl)F
This item is not returnable. View return policy
261762-81-6
C8H5Cl2FO
207.025
MFCD01631371
NWTPJKCARLIKFP-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methylbenzoylchloride, acmc-20alzl, 2-fluoro-6-chloro-3-methyl-benzoyl chloride, 6-chloro-2-fluoro-3-methyl-benzoyl chloride, benzoyl chloride,2-chloro-6-fluoro-3-methyl, 6-chloranyl-2-fluoranyl-3-methyl-benzoyl chloride, benzoyl chloride, 6-chloro-2-fluoro-3-methyl-9ci
2773671
6-chloro-2-fluoro-3-methylbenzoyl chloride
CC1=C(C(=C(C=C1)Cl)C(=O)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.5265
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'