Learn More
6-Bromo-4H-imidazo[4,5-b]pyridine, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 28279-49-4 |
| Molecular Formula | C6H4BrN3 |
| MDL Number | MFCD08706033 |
| Quantity | 1 g |
| SMILES | C1=C(Br)C=NC2=C1[NH]C=N2 |
| Molecular Weight (g/mol) | 198.02 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Bromo-4H-imidazo[4,5-b]pyridine |