Learn More
6-Bromo-2-chloro-1,8-naphthyridine, 96%, Thermo Scientific Chemicals
Product Code. 11444430
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 11444430
Brand: Thermo Scientific Alfa Aesar H50043.03
Request Bulk or Custom Format
6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 902837-40-5
Molecular Formula C8H4BrClN2
Molecular Weight (g/mol) 243.488
MDL Number MFCD07781152
InChI Key KXSBJEOTLVRREX-UHFFFAOYSA-N
Synonym 6-bromo-2-chloro-1,8 naphthyridine, 1,8-naphthyridine, 6-bromo-2-chloro, pubchem18342, ksc495s9r, abbypharma ap-14-10902, 6-bromo-2-chloropyridino 2,3-b pyridine, 6-bromanyl-2-chloranyl-1,8-naphthyridine
PubChem CID 25631443
IUPAC Name 6-bromo-2-chloro-1,8-naphthyridine
SMILES C1=CC(=NC2=NC=C(C=C21)Br)Cl
Specifications
Quantity 1 g
Formula Weight 243.49
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-chloro-1,8-naphthyridine