Learn More
6-Bromo-2,3-difluorotoluene, 95%, Thermo Scientific Chemicals
Catalog Number 15465727
Click to view available options
Quantity:
1 g
5 g
847502-81-2
C7H5BrF2
207.018
MFCD07777161
YXTCBTSPHSNDHD-UHFFFAOYSA-N
12137677
1-bromo-3,4-difluoro-2-methylbenzene
CC1=C(C=CC(=C1F)F)Br
847502-81-2
C7H5BrF2
207.018
MFCD07777161
YXTCBTSPHSNDHD-UHFFFAOYSA-N
12137677
1-bromo-3,4-difluoro-2-methylbenzene
CC1=C(C=CC(=C1F)F)Br
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Refractive Index 1.5105
Quantity 1 g
Synonym 6-bromo-2,3-difluorotoluene, 3,4-difluoro-2-methylbromobenzene, benzene,1-bromo-3,4-difluoro-2-methyl, benzene, 1-bromo-3,4-difluoro-2-methyl, 2-bromo-5,6-difluorotoluene, 3,4-difluoro-2-methyl bromobenzene, 3,4-difluoro-2-methyl-bromobenzene
Formula Weight 207.02
Percent Purity 97%
Chemical Name or Material 6-Bromo-2,3-difluorotoluene