Learn More
6-Azidohexan-1-amine (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | 6-Azidohexan-1-amine |
| CAS | 349553-73-7 |
| Molecular Formula | C6H14N4 |
| Quantity | 100 mg |
| Synonym | 6-Azido-1-hexanamine, 1-Amino-6-azidohexane, 6-Azidohex-1-ylamine, 6-Azidohexylamine |
| InChI Formula | InChI=1S/C6H14N4/c7-5-3-1-2-4-6-9-10-8/h1-7H2 |
| SMILES | NCCCCCCN=[N+]=[N-] |
| IUPAC Name | 6-azidohexan-1-amine |
| Molecular Weight (g/mol) | 142.2 |
| Formula Weight | 142.1218 |