Learn More
(6-Aminohexyl)trimethylammonium Bromide Hydrobromide, TRC
Product Code. 30132858
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132858 250 mg 250mg
30012144 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30132858 Supplier Toronto Research Chemicals Supplier No. TRCA611080250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33968-67-1
Molecular Formula C9 H23 N2 . Br . Br H
Molecular Weight (g/mol) 320.11
Synonym 6-Amino-N,N,N-trimethyl-1-hexanaminium Bromide Hydrobromide, Aminoquat, Colesevelam Aminoquat Impurity HBr
IUPAC Name 6-aminohexyl(trimethyl)azanium;bromide;hydrobromide
SMILES Br.[Br-].C[N+](C)(C)CCCCCCN
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C9H23N2.2BrH/c1-11(2,3)9-7-5-4-6-8-10;;/h4-10H2,1-3H3;2*1H/q+1;;/p-1
Formula Weight 318.0306
Percent Purity >95%
Chemical Name or Material (6-Aminohexyl)trimethylammonium Bromide Hydrobromide
Grade HPLC