Learn More
(6-Amino-3-pyridinyl)methanol, 97%, Thermo Scientific™
Product Code. p-4510262
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 10094962
Brand: Thermo Scientific Maybridge MO07396CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 113293-71-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
InChI Key TXPRFSOGPYITOT-UHFFFAOYSA-N
PubChem CID 2794829
IUPAC Name (6-aminopyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)N
Specifications
Assay Percent Range 97%
Quantity 250 mg
Synonym 6-aminopyridin-3-yl methanol, 6-amino-3-pyridinyl methanol, 6-amino-3-pyridinemethanol, 2-amino-5-hydroxymethylpyridine, 6-amino-pyridin-3-yl-methanol, 2-amino-5-hydroxymethyl pyridine, 2-amino-5-pyridinemethanol, 3-pyridinemethanol, 6-amino, 3-pyridinemethanol,6-amino, 6-amino-3-hydroxymethylpyridine
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material (6-Amino-3-pyridinyl)methanol