Learn More
6-Amino-3-chloropicolinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Amino-3-chloropicolinic Acid |
| CAS | 1020917-27-4 |
| Molecular Formula | C25 H32 O12 |
| Quantity | 250 mg |
| Synonym | 6-Amino-3-chloro-2-pyridinecarboxylic Acid |
| InChI Formula | InChI=1S/C25H32O12/c1-11(7-9-15(26)32-3)6-8-13-20(33-4)12(2)14-10-35-23(30)16(14)21(13)36-25-19(29)17(27)18(28)22(37-25)24(31)34-5/h6,17-19,22,25,27-29H,7-10H2,1-5H3/b11-6+/t17?,18-,19?,22?,25-/m1/s1 |
| SMILES | COC(=O)CC\C(=C\Cc1c(OC)c(C)c2COC(=O)c2c1O[C@@H]3OC([C@H](O)C(O)C3O)C(=O)OC)\C |
| IUPAC Name | methyl (3R,6S)-3,4,5-trihydroxy-6-[[6-methoxy-5-[(E)-6-methoxy-3-methyl-6-oxohex-2-enyl]-7-methyl-3-oxo-1H-2-benzofuran-4-yl]oxy]oxane-2-carboxylate |
| Molecular Weight (g/mol) | 524.514 |
| Formula Weight | 524.189 |