missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methylquinoline, 97%, Thermo Scientific Chemicals

Catalog Number 15463077
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
65079-19-8
C10H10N2
158.204
MFCD00052600
TYJFYUVDUUACKX-UHFFFAOYSA-N
103148
2-methylquinolin-6-amine
CC1=NC2=C(C=C1)C=C(C=C2)N
This item is not returnable. View return policy
65079-19-8
C10H10N2
158.204
MFCD00052600
TYJFYUVDUUACKX-UHFFFAOYSA-N
103148
2-methylquinolin-6-amine
CC1=NC2=C(C=C1)C=C(C=C2)N

6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 189°C to 191°C
Quantity 250 mg
Beilstein 114795
Synonym 6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine
Solubility Information Soluble in water.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 6-Amino-2-methylquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'