missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-1-methyluracil, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.
Solubility
Soluble in diluted sodium hydroxide solution.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.
Solubility
Soluble in diluted sodium hydroxide solution.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | >300°C |
Beilstein | 127771 |
Solubility Information | Soluble in diluted sodium hydroxide solution. |
Formula Weight | 141.13 |
Percent Purity | 97% |
Quantity | 25 g |
Odor | Odorless |
Chemical Name or Material | 6-Amino-1-methyluracil |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'