missing translation for 'onlineSavingsMsg'
Learn More

6-Alpha-Methylprednisolone Crystalline MP Biomedicals

Product Code. 11432412 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432412 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432412 Supplier MP Biomedicals Supplier No. 0215167191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 83-43-2
Molecular Formula C22H30O5
MDL Number MFCD00010591
Quantity 1 g
Synonym methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol
InChI Key VHRSUDSXCMQTMA-PJHHCJLFSA-N
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12
IUPAC Name (1R,3aS,3bS,5S,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-5,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol) 374.48
PubChem CID 6741
ChEBI CHEBI:6888
Formula Weight 374.5
Physical Form Crystalline
Chemical Name or Material 6-Alpha-Methylprednisolone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.