missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 11476310
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476310 1 g 1g
15497655 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476310 Supplier Thermo Scientific Alfa Aesar Supplier No. H27168.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in organic synthesis. The boronic esters are widely used for carrying out cross coupling reactions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-81-0
Molecular Formula C17H19BFNO2
Molecular Weight (g/mol) 299.15
MDL Number MFCD09037494
InChI Key CTCLIEHCPOJNSW-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-4-fluorophenyl pyridine-3-boronic acid pinacol ester, 2-4-fluorophenyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-4-fluorophenyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 44755190
IUPAC Name 2-(4-fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C1=CC=C(F)C=C1

Specifications

Quantity 1 g
Formula Weight 299.15
Percent Purity 95%
Chemical Name or Material 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.