Learn More
6-(1-Pyrrolidinyl)nicotinaldehyde, Thermo Scientific™
Catalog Number p-4510207
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
261715-39-3
C10H12N2O
176.219
FFFAXGTZBUMKTI-UHFFFAOYSA-N
6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
2794775
6-pyrrolidin-1-ylpyridine-3-carbaldehyde
C1CCN(C1)C2=NC=C(C=C2)C=O
261715-39-3
C10H12N2O
176.219
FFFAXGTZBUMKTI-UHFFFAOYSA-N
6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
2794775
6-pyrrolidin-1-ylpyridine-3-carbaldehyde
C1CCN(C1)C2=NC=C(C=C2)C=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 6-(1-Pyrrolidinyl)nicotinaldehyde