missing translation for 'onlineSavingsMsg'
Learn More

YHO-13177, MedChemExpress

Product Code. 16448037
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448037 5 mg 5mg
16495318 10 mg 10mg
16485966 100 mg 100mg
16441527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448037 Supplier Medchem Express Supplier No. HY12757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YHO-13177 is a potent and specific inhibitor of BCRP; potentiated the cytotoxicity of SN-38 in cancer cells and no effect on P-glycoprotein-mediated paclitaxel resistance in MDR1-transduced human leukemia K562 cells.

YHO-13177 is a potent and specific inhibitor of BCRP

Specifications

Chemical Name or Material YHO-13177
CAS 912287-56-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H22N2O3S
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (89.97 mM; Need ultrasonic)
SMILES N#C/C(C1=CC=C(OC)C(OC)=C1)=C\C2=CC=C(N3CCC(O)CC3)S2
Molecular Weight (g/mol) 370.47
Formula Weight 370.47
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.