missing translation for 'onlineSavingsMsg'
Learn More

Vipadenant, MedChemExpress

Product Code. 16426927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426927

Brand: Medchem Express HY10857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vipadenant (BIIB-014; CEB-4520) is an adenosine receptor antagonist, with Kis of 1.3 nM and 68 nM for A2A and A1, respectively.

Vipadenant (BIIB-014

Specifications

Chemical Name or Material Vipadenant
Physical Form Solid
CAS 442908-10-3
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H15N7O
For Use With (Application) Neuroscience-Neuromodulation
Synonym BIIB-014 CEB-4520
Solubility Information DMSO : ≥ 31 mg/mL (96.47 mM)
SMILES NC1=NC2=C(N=NN2CC3=CC=C(N)C(C)=C3)C(C4=CC=CO4)=N1
Molecular Weight (g/mol) 321.34
Formula Weight 321.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.