missing translation for 'onlineSavingsMsg'
Learn More

Vilanterol trifenatate, MedChemExpress

Product Code. 16448637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16448637 5 mg 5mg
16482167 50 mg 50mg
16476028 10 mg 10mg
16416446 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16448637

Brand: Medchem Express HY14300A5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vilanterol trifenatate (GW642444 trifenatate) is a long-acting β2-adrenoceptor (β2-AR) agonist with inherent 24-hour activity. The pEC50s for β2-AR, β1-AR and β3-AR are 10.37, 6.98 and 7.36, respectively.

Vilanterol trifenatate (GW642444 trifenatate) is a long-acting β2-adrenoceptor (β2-AR) agonist with inherent 24-hour activity. The pEC50s for β2-AR, β1-AR and β3-AR are 10.37, 6.98 and 7.36, respectively.

Specifications

Chemical Name or Material Vilanterol trifenatate
CAS 503070-58-4
Color White
Purity Grade Notes Research
Molecular Formula C44H49Cl2NO7
Quantity 5 mg
Synonym GW642444 trifenatate
Solubility Information DMSO : 100 mg/mL (129.07 mM; Need ultrasonic)
SMILES OC1=CC=C([C@@H](O)CNCCCCCCOCCOCC2=C(Cl)C=CC=C2Cl)C=C1CO.O=C(O)C(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5
Molecular Weight (g/mol) 774.77
Formula Weight 774.77
Percent Purity 97.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.