missing translation for 'onlineSavingsMsg'
Learn More

TRC051384, MedChemExpress

Product Code. 16446247
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446247 5 mg 5mg
16493518 10 mg 10mg
16460936 200 mg 200mg
16414666 100 mg 100mg
16409966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16446247 Supplier Medchem Express Supplier No. HY101712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TRC051384 is a heat shock protein 70 (HSP70) inducer.

TRC051384 is a heat shock protein 70 (HSP70) inducer.

Specifications

Chemical Name or Material TRC051384
CAS 867164-40-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H31N5O4
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (214.80 mM)
SMILES O=C(NC1=CC=C(C(/C=C/C2=NC(N3CCOCC3)=CC=C2)=O)C=C1)NCCN4CCOCC4
Molecular Weight (g/mol) 465.54
Formula Weight 465.54
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.