missing translation for 'onlineSavingsMsg'
Learn More

TPA 023, MedChemExpress

Product Code. 16486197
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. Quantity unitSize
16486197 5 mg 5mg
16440829 1 mg 1mg
16433478 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16486197

Brand: Medchem Express HY101640

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TPA 023 is a GABAA α2/α3 subtype-selective agonist, with Ki of 0.19-0.41 nM.

TPA 023 is a GABAA α2/α3 subtype-selective agonist, with Ki of 0.19-0.41 nM.

Specifications

Chemical Name or Material TPA 023
CAS 252977-51-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22FN7O
Quantity 5 mg
SMILES FC1=CC=CC=C1C2=NN=C3C=C(C(C)(C)C)C(OCC4=NC=NN4CC)=NN32
Molecular Weight (g/mol) 395.43
Formula Weight 395.43
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.