missing translation for 'onlineSavingsMsg'
Learn More

CCR2 antagonist 4, MedChemExpress

Product Code. 16456897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16456897

Brand: Medchem Express HY108323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CCR2 antagonist 4 (Teijin compound 1) is a potent and specific CCR2 antagonist, with IC50s of 180 nM for CCR2b. CCR2 antagonist 4 potently inhibits MCP-1-induced chemotaxis with an IC50 of 24 nM.

CCR2 antagonist 4 (Teijin compound 1) is a potent and specific CCR2 antagonist, with IC50s of 180 nM for CCR2b. CCR2 antagonist 4 potently inhibits MCP-1-induced chemotaxis with an IC50 of 24 nM.

Specifications

Chemical Name or Material CCR2 antagonist 4
Physical Form Powder
CAS 226226-39-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H21ClF3N3O2
For Use With (Application) Cancer-programmed cell death
Synonym Teijin compound 1
Solubility Information DMSO : ≥ 50 mg/mL (113.67 mM)
SMILES O=C(NCC(N[C@H]1CN(CC2=CC=C(Cl)C=C2)CC1)=O)C3=CC=CC(C(F)(F)F)=C3
Molecular Weight (g/mol) 439.86
Formula Weight 439.86
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.