missing translation for 'onlineSavingsMsg'
Learn More

SYP-5, MedChemExpress

Product Code. 16465857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465857

Brand: Medchem Express HY100693

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SYP-5 is a novel HIF-1 inhibitor, suppresses tumor cells invasion and angiogenesis.

SYP-5 is a novel HIF-1 inhibitor, suppresses tumor cells invasion and angiogenesis.

Specifications

Chemical Name or Material SYP-5
CAS 1384268-04-5
Color Orange
Purity Grade Notes Research
Molecular Formula C18H16O3S
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (106.70 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C(C=CC(C)(C)O2)=C2C=C1)/C=C/C3=CC=CS3
Molecular Weight (g/mol) 312.38
Formula Weight 312.38
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.