missing translation for 'onlineSavingsMsg'
Learn More

SR1078, MedChemExpress

Product Code. 16448667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16448667 5 mg 5mg
16486058 10 mg 10mg
16472197 50 mg 50mg
16406466 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16448667

Brand: Medchem Express HY14422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription.

SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription.

Specifications

Chemical Name or Material SR1078
Physical Form Solid
CAS 1246525-60-9
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H10F9NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 33.33 mg/mL (77.29 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(C(F)(F)F)(C(F)(F)F)O)C=C1)C2=CC=C(C(F)(F)F)C=C2
Molecular Weight (g/mol) 431.25
Formula Weight 431.25
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.