missing translation for 'onlineSavingsMsg'
Learn More

SKF-86002, MedChemExpress

Product Code. 16457947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457947 5 mg 5mg
16465228 10 mg 10mg
16401447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457947 Supplier Medchem Express Supplier No. HY12511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKF-86002 is an orally active p38 MAPK inhibitor, with anti-inflammatory, anti-arthritic and analgesic activities. SKF-86002 inhibits lipopolysaccharide (LPS)-stimulate human monocyte IL-1 and TNF-α production (IC50 = 1 μM). SKF-86002 inhibits lipoxygenase- and cyclooxygenase-mediated metabolism of arachidonic acid.

SKF-86002 is an orally active p38 MAPK inhibitor, with anti-inflammatory, anti-arthritic and analgesic activities. SKF-86002 inhibits lipopolysaccharide (LPS)-stimulate human monocyte IL-1 and TNF-α production (IC50 = 1 μM). SKF-86002 inhibits lipoxygenase- and cyclooxygenase-mediated metabolism of arachidonic acid.

Specifications

Chemical Name or Material SKF-86002
CAS 72873-74-6
Color Brown
Purity Grade Notes Research
Molecular Formula C16H12FN3S
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (112.09 mM; Need ultrasonic)
SMILES FC1=CC=C(C2=C(C3=CC=NC=C3)N4C(SCC4)=N2)C=C1
Molecular Weight (g/mol) 297.35
Formula Weight 297.35
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.