missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWJ 50271, MedChemExpress
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
RWJ 50271 is an selective and orally active inhibitor of lymphocyte function-associated antigen-1/intercellular adhesion molecule-1(LFA-1/ICAM-1) interaction with an IC50 of 5.0 μM (HL60 cells). RWJ 50271 inhibits LFA-1/ICAM-1-mediated cell adhesion.
Specifications
Specifications
| Chemical Name or Material | RWJ 50271 |
| Physical Form | Solid |
| CAS | 162112-37-0 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H17F3N4O2S |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 250 mg/mL (609.15 mM; Need ultrasonic) |
| SMILES | CC1=C(C(NCCCO)=O)C=NN1C2=NC(C3=CC(C(F)(F)F)=CC=C3)=CS2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction