missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pritelivir, MedChemExpress
Description
Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.
Specifications
Specifications
| Chemical Name or Material | Pritelivir |
| Physical Form | Powder |
| CAS | 348086-71-5 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18N4O3S2 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | AIC316 BAY 57-1293 |
| Solubility Information | DMSO : ≥ 33 mg/mL (81.99 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?