missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF 1022A, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
PF 1022A is a cyclooctadepsipeptide with broadspectrum anthelmintic properties produced by fermentation of the fungus Mycelia sterilia. PF 1022A is a channel-forming ionophore. PF 1022A showes strong anthelmintic activities against Ascaridia galli in chickens. PF 1022A also can be used for angiostrongyliasis research.
Specifications
Specifications
| Chemical Name or Material | PF 1022A |
| Physical Form | Solid |
| CAS | 133413-70-4 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C52H76N4O12 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 43 mg/mL (45.30 mM) |
| SMILES | O=C(N([C@H](C(O[C@](C(N([C@H](C(O[C@H](CC1=CC=CC=C1)C2=O)=O)CC(C)C)C)=O)([H])C)=O)CC(C)C)C)[C@H](OC([C@@H](N(C([C@H](OC([C@](N2C)([H])CC(C)C)=O)C)=O)C)CC(C)C)=O)CC3=CC=CC=C3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction