missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pasireotide ditrifluoroacetate, MedChemExpress
Description
Pasireotide (SOM230) ditrifluoroacetate, a long-acting cyclohexapeptide somatostatin analogue, can improve agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5, pKi=8.2/9.0/9.1/<7.0/9.9, respectively). Pasireotide ditrifluoroacetate exhibits antisecretory, antiproliferative, and proapoptotic activity.
Specifications
Specifications
| Chemical Name or Material | Pasireotide ditrifluoroacetate |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C62H68F6N10O13 |
| Quantity | 5 mg |
| Synonym | SOM230 ditrifluoroacetate Pasireotide TFA salt |
| Solubility Information | DMSO : 100 mg/mL (78.42 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (26.14 mM; Need ultrasonic) |
| SMILES | O=C(O)C(F)(F)F.O=C(O)C(F)(F)F.NCCCC[C@@H](C(N[C@H]1CC2=CC=C(C=C2)OCC3=CC=CC=C3)=O)NC([C@H](NC([C@H](C4=CC=CC=C4)NC([C@H]5N(C([C@H](CC6=CC=CC=C6)NC1=O)=O)C[C@H](OC(NCCN)=O)C5)=O)=O)CC7=CNC8=C7C=CC=C8)=O |
| Molecular Weight (g/mol) | 1275.25 |
| Formula Weight | 1275.25 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?