missing translation for 'onlineSavingsMsg'
Learn More

Loureirin A, MedChemExpress

Product Code. 16481648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
16481648 5 mg 5mg
1 options
This item is not returnable. View return policy

Product Code. 16481648

Brand: Medchem Express HYN1505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loureirin A is a flavonoid extracted from Dragon's Blood, can inhibit Akt phosphorylation, and has antiplatelet activity.

Loureirin A is a flavonoid extracted from Dragon's Blood, can inhibit Akt phosphorylation, and has antiplatelet activity.

Specifications

Chemical Name or Material Loureirin A
CAS 119425-89-7
Color White
Purity Grade Notes Research
Molecular Formula C17H18O4
Quantity 5 mg
Solubility Information DMSO : ≥ 86.6 mg/mL (302.46 mM)
SMILES O=C(C1=CC=C(O)C=C1)CCC2=CC=C(OC)C=C2OC
Molecular Weight (g/mol) 286.32
Formula Weight 286.32
Percent Purity 98.93%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.