missing translation for 'onlineSavingsMsg'
Learn More

LGD-6972, MedChemExpress

Product Code. 16457957
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457957

Brand: Medchem Express HY12525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LGD-6972 is a selective and orally active glucagon receptor antagonist. LGD-6972 has the potential for type 2 diabetes research.

LGD-6972 is a selective and orally active glucagon receptor antagonist. LGD-6972 has the potential for type 2 diabetes research.

Specifications

Chemical Name or Material LGD-6972
CAS 1207989-09-0
Color White
Purity Grade Notes Research
Molecular Formula C43H46N2O5S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (177.83 mM; Need ultrasonic)
SMILES O=C(NCCS(=O)(O)=O)C1=CC=C(C[C@H](C2=CC=C(C3=CC=C(C(C)(C)C)C=C3)C=C2)C(NC4=CC=C(C5=C(C)C=C(C)C=C5C)C=C4)=O)C=C1
Molecular Weight (g/mol) 702.9
Formula Weight 702.9
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.